Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33325

Sigma-Aldrich

Potassium phthalate monobasic

puriss. p.a., buffer substance, ≥99.5%

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein/REAXYS Number:
3637128
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

assay

≥99.5%

form

solid

loss

≤0.2% loss on drying, 105 °C

pH

4

mp

295-300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phthalate monobasic is the monopotassium salt of phthalic acid. It has been proposed as a certified reference material (CRM) under ISO/IEC 17025 and ISO Guide 34 accreditation. Its spectral shift, multiplicity, solubility and relaxation times in different solvents have been reported.[1]

Application

Potassium phthalate monobasic may be used as a buffer to maintain pH = 2.39 and 4.52, during the dispersive ionic liquid-liquid microextraction coupled with liquid chromatography and UV detection of drug (benznidazole and nifurtimox) levels in human plasma.[2] It has been used as internal standard certified reference material (CRM) for the high performance (HP)-quantitative NMR spectroscopic (qNMR) determination of various organic compounds.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

387.1 °F - Pensky-Martens closed cup

flash_point_c

197.3 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M Padró et al.
Talanta, 107, 95-102 (2013-04-20)
Dispersive ionic liquid-liquid microextraction combined with liquid chromatography and UV detection was used for the determination of two antichagasic drugs in human plasma: nifurtimox and benznidazole. The effects of experimental parameters on extraction efficiency-the type and volume of ionic liquid
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional
Martha M Téllez-Rojo et al.
The Science of the total environment, 461-462, 386-390 (2013-06-12)
Previous studies suggest that prenatal phthalate exposure affects neurodevelopment and behavior during the first years of life. To evaluate the effect of maternal urinary concentrations of phthalate metabolites during pregnancy on mental and psychomotor development in children 24-36 months of
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Hanne Frederiksen et al.
Environmental science & technology, 47(2), 958-967 (2012-12-14)
Urinary phthalate excretion is used as marker of phthalate exposure in epidemiological studies. Here we examine the reliability of urinary phthalate levels in exposure classification by comparing the inter- and intrasubject variation of urinary phthalate metabolite levels. Thirty-three young healthy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service