32916
Fumonisin B3-13C34 solution
~10 μg/mL in acetonitrile: water, analytical standard
About This Item
Recommended Products
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
~10 μg/mL in acetonitrile: water
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
single component solution
mass shift
M+34
storage temp.
−20°C
SMILES string
[13CH3][13CH2][13CH2][13CH2][13C@@H]([13CH3])[13C@@H](O[13C](=O)[13CH2][13CH]([13CH2][13C](O)=O)[13C](O)=O)[13C@H]([13CH2][13C@@H]([13CH3])[13CH2][13C@H](O)[13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C@H](O)[13C@@H]([13CH3])N)O[13C](=O)[13CH2][13CH]([13CH2][13C](O)=O)[13C](O)=O
InChI
1S/C34H59NO14/c1-5-6-11-21(3)32(49-31(43)19-24(34(46)47)17-29(40)41)27(48-30(42)18-23(33(44)45)16-28(38)39)15-20(2)14-25(36)12-9-7-8-10-13-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)/t20-,21+,22+,23?,24?,25+,26-,27-,32+/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1,19+1,20+1,21+1,22+1,23+1,24+1,25+1,26+1,27+1,28+1,29+1,30+1,31+1,32+1,33+1,34+1
InChI key
CPCRJSQNWHCGOP-BMFHXWBQSA-N
General description
Application
Analysis Note
related product
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
42.8 °F
flash_point_c
6 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service