Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32888

Supelco

Nonylphenol solution

50 μg/mL in acetone, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

−20°C

InChI

1S/C15H24O/c1-13(2)7-5-3-4-6-8-14-9-11-15(16)12-10-14/h9-13,16H,3-8H2,1-2H3

InChI key

JSFITYFUKSFPBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonylphenol belongs to the class of alkylphenol compounds produced from the biodegradation of non-ionic surfactants alkylphenol ethoxylates (APEOs) primarily utilized as detergents, emulsifiers and lubricants.[1]

Application

Nonylphenol may be used as an analytical reference standard for the determination of the analyte in aqueous samples using simultaneous derivatization and dispersive liquid–liquid microextraction (DLLME) followed by gas chromatography–mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shusheng Luo et al.
Journal of chromatography. A, 1217(43), 6762-6768 (2010-07-10)
A simple and fast sample preparation method for the determination of nonylphenol (NP) and octylphenol (OP) in aqueous samples by simultaneous derivatization and dispersive liquid-liquid microextraction (DLLME) was investigated using gas chromatography-mass spectrometry (GC/MS). In this method, a combined dispersant/derivatization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service