Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32881

Supelco

4-tert-Octylphenol solution

1 μg/mL in acetone, analytical standard

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

1 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(O)cc1

InChI

1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3

InChI key

ISAVYTVYFVQUDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Hui Li
Environmental toxicology and chemistry, 31(4), 843-850 (2012-01-27)
Surfactants are a major class of emerging pollutants widely used in large quantities in everyday life and commonly found in surface waters worldwide. Freshwater planarian was selected to examine the effects of different surfactants by measuring mortality, mobility, and membrane-bound
Tadashi Toyama et al.
Environmental science & technology, 45(15), 6524-6530 (2011-07-09)
We investigated biodegradation of technical nonylphenol (tNP) in Phragmites australis rhizosphere sediment by conducting degradation experiments using sediments spiked with tNP. Accelerated tNP removal was observed in P. australis rhizosphere sediment, whereas tNP persisted in unvegetated sediment without plants and
Qingxiang Zhou et al.
Talanta, 85(3), 1598-1602 (2011-08-03)
Present study described a simple, sensitive, and viable method for the determination of bisphenol A, 4-n-nonylphenol and 4-tert-octylphenol in water samples using temperature-controlled ionic liquid dispersive liquid-phase microextraction coupled to high performance liquid chromatography-fluorescence detector. In this experiment, 1-octyl-3-methylimidazolium hexafluorophosphate
Wen-Ling Chen et al.
Talanta, 89, 237-245 (2012-01-31)
This study developed and validated a method of measuring the feminizing chemicals 4-tert-octylphenol, 4-nonylphenol, nonylphenol monoethoxycarboxylate (NP(1)EC), nonylphenol monoethoxylate (NP(1)EO), nonylphenol diethoxylate (NP(2)EO), estrone, 17β-estradiol, estriol, 17α-ethinyl estradiol and bisphenol A in river water, sediment, and tissue using ultra-high performance
Jian Gong et al.
Environmental toxicology and chemistry, 31(11), 2456-2464 (2012-08-01)
The distribution of endocrine-disrupting chemicals (EDCs) and its relationship with dissolved and particulate organic carbon (DOC and POC) was investigated in selected rivers of the Pearl River Delta, South China. The aqueous concentrations (average; ng/L) and particulate concentrations (average; ng/g

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service