Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32843

Supelco

Hydroxymebendazole-d3

VETRANAL®, analytical standard

Synonym(s):

[5-(Hydroxyphenylmethyl)-2-benzimidazolecarbamic acid methyl-d3 ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H12N3O3
CAS Number:
Molecular Weight:
300.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)Nc1nc2cc(ccc2[nH]1)C(O)c3ccccc3

InChI

1S/C16H15N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9,14,20H,1H3,(H2,17,18,19,21)/i1D3

InChI key

IIQKUGXEGMZCLE-FIBGUPNXSA-N

General description

Hydroxymebendazole-d3 is the deuterated compound of anthelmintic drug, hydroxymebendazole.[1]

application

Hydroxymebendazole-d3 may be used as an internal standard for the determination of hydroxymebendazole in milk samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of anthelmintic drug residues in milk using ultra high performance liquid chromatography-tandem mass spectrometry with rapid polarity switching.
Whelan M, et al.
Journal of Chromatography A, 1217(27), 4612-4622 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service