Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32817

Supelco

EPA 624 Calibration Mix 1

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

packaging

ampule of 2 mL

concentration

100 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-8°C

General description

Certan Vial
for EU only, for US: Supelco 47506

EPA 624 Calibration Mix 1 can be utilized for identifying and quantifying purgeable halocarbons in municipal and industrial wastewater. The pre-requisite of EPA 624 method is the use of gas chromatography with a selective detector or for gas chromatography/mass spectrometry. The analyst may be better equipped to choose between GC and HPLC.
Packed GC columns are described in the EPA 624 method, but chromatographic conditions can be changed (e.g., capillary columns may be used) if the changes do not make the method "less accurate or less precise" than the standard method.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service