Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32811

Supelco

Monosultap

PESTANAL®, analytical standard

Synonym(s):

Monomehypo, Thiosulfuric acid S,S′-[2-(dimethylamino)trimethylene] ester monosodium salt, Thiosultap monosodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12NNaO6S4
CAS Number:
Molecular Weight:
333.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C(CSS(O)(=O)=O)CSS(=O)(=O)O[Na]

InChI

1S/C5H13NO6S4.Na/c1-6(2)5(3-13-15(7,8)9)4-14-16(10,11)12;/h5H,3-4H2,1-2H3,(H,7,8,9)(H,10,11,12);/q;+1/p-1

InChI key

MBNMHBAJUNHZRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Monosultap is one of the nereistoxin derivatives that is known as a stomach poison.[1]

Application

Monosultap may be used as an analytical reference standard for the determination of the analyte in tomato and soil samples by capillary gas chromatography with flame photometric detection.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Antidotal effect of sodium dimercapto-propane sulphonate in the treatment of acute poisoning of 2-dimethylamino-1,3-histhio-sulfopropanedisodium salt in 20 cases].
Y F Di
Zhonghua nei ke za zhi, 32(12), 827-827 (1993-12-01)
Fengzu Zhang et al.
Bulletin of environmental contamination and toxicology, 88(3), 362-367 (2011-10-29)
A modified method for the analysis of monosultap residue in rice plant and environment was developed and validated. Monosultap residue dynamics and final residues in supervised field trials at GAP conditions were studied. At fortification levels of 0.05, 0.5 and
Z Q Lu et al.
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 26(4), 213-215 (1992-07-01)
In mice, DMPS (250 mg/kg, i.v.) combined with diazepam (1.25 mg/kg, i.p.) could increase LD50 of p. o. SCD 5.3 times. DMPS (62.5 mg/kg, i.v.) antagonized completely the respiratory depression and neuromuscular blockade caused by SCD(7.5 mg/kg, i.v.) in rabbits.
Determination of insecticide monosultap residues in tomato and soil by capillary gas chromatography with flame photometric detection.
Tao CJ, et al.
Canadian Journal of Analytical Sciences and Spectroscopy, 295-295 (2007)
B J Cao et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 11(3), 207-210 (1990-05-01)
The neuromuscular blocking and respiratory depressing actions of the new insecticide sodium ammonium dimethyl-2-(propano-1,3-dithiosulfate) monohydrate (SCD) were investigated. In peroneal-tibialis anterior nerve-muscle preparations of urethane anesthetized rabbit, SCD 6.5 mg/kg iv completely depressed the indirectly elicited twitch tension but not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service