Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32790

Sigma-Aldrich

1,4-Diaminobutane

puriss., ≥99.0% (GC)

Synonym(s):

1,4-Butanediamine, Putrescine, Tetramethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2
CAS Number:
Molecular Weight:
88.15
Beilstein/REAXYS Number:
605282
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (GC)

form

solid

expl. lim.

9.08 %

refractive index

n20/D 1.457 (lit.)
n20/D 1.457

bp

158-160 °C (lit.)

mp

25-28 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

SMILES string

NCCCCN

InChI

1S/C4H12N2/c5-3-1-2-4-6/h1-6H2

InChI key

KIDHWZJUCRJVML-UHFFFAOYSA-N

Gene Information

rat ... Odc1(24609)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

113.0 °F - closed cup

flash_point_c

45 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juhani Jänne et al.
European journal of biochemistry, 271(5), 877-894 (2004-03-11)
The polyamines putrescine, spermidine and spermine are organic cations shown to participate in a bewildering number of cellular reactions, yet their exact functions in intermediary metabolism and specific interactions with cellular components remain largely elusive. Pharmacological interventions have demonstrated convincingly
Barry J Shelp et al.
Plant science : an international journal of experimental plant biology, 193-194, 130-135 (2012-07-17)
4-Aminobutyrate (GABA) accumulates in various plant parts, including bulky fruits such as apples, in response to abiotic stress. It is generally believed that the GABA is derived from glutamate, although a contribution from polyamines is possible. Putrescine, but not spermidine
Chi Yang et al.
PloS one, 7(10), e47314-e47314 (2012-10-17)
The phosphate starvation response in bacteria has been studied extensively for the past few decades and the phosphate-limiting signal is known to be mediated via the PhoBR two-component system. However, the global DNA binding profile of the response regulator PhoB
Lianghua Chen et al.
Ecotoxicology and environmental safety, 97, 94-102 (2013-08-14)
We used the dioecious tree, Populus cathayana, as a model species to study plants' physiological and biochemical responses to copper (Cu) stress, exogenous putrescine (Put) treatment and their interaction. Although males accumulated higher Cu concentrations in leaves than did females
Radika Soysa et al.
The Biochemical journal, 452(3), 423-432 (2013-03-29)
The TcPOT1.1 gene from Trypanosoma cruzi encodes a high affinity putrescine-cadaverine transporter belonging to the APC (amino acid/polyamine/organocation) transporter superfamily. No experimental three-dimensional structure exists for any eukaryotic member of the APC family, and thus the structural determinants critical for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service