Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

327751

Supelco

Polystyrene

analytical standard, for GPC, 35,000

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard
for GPC

Quality Level

form

powder

mol wt

average Mw 35,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene (for GPC, 35,000) is a synthetic thermoplastic, which has a broad range of applications on account of its properties such as cheap cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weathering of Polymers (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service