Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C17H19N5O6S
CAS Number:
Molecular Weight:
421.43
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
9011949
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
COc1cc(OC)nc(NC(=O)NS(=O)(=O)Nc2ccccc2C(=O)C3CC3)n1
InChI
1S/C17H19N5O6S/c1-27-13-9-14(28-2)19-16(18-13)20-17(24)22-29(25,26)21-12-6-4-3-5-11(12)15(23)10-7-8-10/h3-6,9-10,21H,7-8H2,1-2H3,(H2,18,19,20,22,24)
General description
Cyclosulfamurons belong to the class of pyrimidinylsulfonylurea herbicides that are used for weed control on a variety of agricultural products.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Cyclosulfamuron may be used as an analytical reference standard for the determination of the analyte in:
- Cereal grain samples by ultra-performance liquid chromatography–electrospray ionization-mass spectrometry (UPLC-ESI-MS).
- Shellfish by LC coupled to tandem mass spectrometry (MS/MS).
Other Notes
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Development of multi-residue analysis of herbicides in cereal grain by ultra-performance liquid chromatography-electrospray ionization-mass spectrometry.
Dong X, et al.
Food Chemistry, 192(23), 432-440 (2016)
Study and determination of the herbicide cyclosulfamuron by square wave stripping voltammetry.
Sar?gul T and ?nam, R
Electrochimica Acta, 54(23), 5376-5380 (2009)
Analysis of banned veterinary drugs and herbicide residues in shellfish by liquid chromatography-tandem mass spectrometry (LC/MS/MS) and gas chromatography-tandem mass spectrometry (GC/MS/MS).
Chang GR, et al.
Marine Pollution Bulletin, 113(1-2), 579-584 (2016)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SML3877-5MG | 04065270979940 |
| SML3877-25MG | 04065270979933 |
| 32743-100MG | 04061826716816 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service