Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32725

Supelco

Imibenconazole solution

PESTANAL®, analytical standard

Synonym(s):

4-Chlorobenzyl N-(2,4-dichlorophenyl)-1H-1,2,4-triazole-1-acetimidothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13Cl3N4S
CAS Number:
Molecular Weight:
411.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(CS\C(Cn2cncn2)=N\c3ccc(Cl)cc3Cl)cc1

InChI

1S/C17H13Cl3N4S/c18-13-3-1-12(2-4-13)9-25-17(8-24-11-21-10-22-24)23-16-6-5-14(19)7-15(16)20/h1-7,10-11H,8-9H2/b23-17+

InChI key

AGKSTYPVMZODRV-HAVVHWLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imibenconazoles belong to the family of azole-type fungicides.[1]

Application

Imibenconazole may be used as an analytical reference standard for the determination of the analyte in:
  • Honeybees by acetonitrile-based extraction and gas chromatography–tandem quadrupole mass spectrometry (GC-MS/MS).[2]
  • Agricultural products by acetonitrile-based extraction and matrix solid-phase dispersion (MSPD), followed by online gel permeation chromatography (GPC) combined with GC/MS.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

39.2 °F - closed cup

flash_point_c

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of automated online gel permeation chromatography-gas chromatograph mass spectrometry for measuring multiresidual pesticides in agricultural products.
Liu LB, et al.
Journal of Chromatography A, 845(1), 61-68 (2007)
Inhibitory effects of azole-type fungicides on interleukin-17 gene expression via retinoic acid receptor-related orphan receptors α and γ
Kojima H, et al.
Toxicology and Applied Pharmacology, 259(3), 338-345 (2012)
Development and validation of a multi-residue method for the determination of pesticides in honeybees using acetonitrile-based extraction and gas chromatography-tandem quadrupole mass spectrometry
Walorczyk S and Gnusowski B
Journal of Chromatography A, 1216(37), 6522-6531 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service