Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32648

Supelco

S-Metolachlor Metabolite CGA 50267

PESTANAL®, analytical standard

Synonym(s):

(S)-2-(2-Ethyl-6-methylphenylamino)propionic acid, N-(2-Ethyl-6-methylphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
Molecular Weight:
207.27
Beilstein/REAXYS Number:
6481181
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

SMILES string

CCc1cccc(C)c1N[C@@H](C)C(O)=O

InChI

1S/C12H17NO2/c1-4-10-7-5-6-8(2)11(10)13-9(3)12(14)15/h5-7,9,13H,4H2,1-3H3,(H,14,15)/t9-/m0/s1

InChI key

ACQCQKSFTBFYDO-VIFPVBQESA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service