Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32599

Supelco

Dapsone-13C12

VETRANAL®, analytical standard

Synonym(s):

4,4′-Diaminodiphenyl-13C12 sulfone, 4,4′-Sulfonyldianiline-13C12, Bis(4-aminophenyl-13C6) sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C12H12N2O2S
CAS Number:
Molecular Weight:
260.21
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 1B - STOT RE 2 - STOT SE 2

target_organs

Blood, Blood,spleen,Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service