Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32580

Supelco

Clenhexerol

VETRANAL®, analytical standard

Synonym(s):

1-(4-Amino-3,5-dichloro-phenyl)-2-hexylaminoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22Cl2N2O
CAS Number:
Molecular Weight:
305.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCNCC(O)c1cc(Cl)c(N)c(Cl)c1

InChI

1S/C14H22Cl2N2O/c1-2-3-4-5-6-18-9-13(19)10-7-11(15)14(17)12(16)8-10/h7-8,13,18-19H,2-6,9,17H2,1H3

InChI key

OIJJTCCRNKYZHW-UHFFFAOYSA-N

General description

Clenhexerol is classified under the β-agonist group of compounds.[1]

Application

Clenhexerol may be used as an analytical reference standard for the determination of the analyte in:
  • Animal feed samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and liquid chromatography-high resolution mass spectrometry (LC-HRMS).[2]
  • Bovine urine samples by QuEChERS extraction and ultra-high performance LC combined with HRMS.[1]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wide-range screening of banned veterinary drugs in urine by ultra high liquid chromatography coupled to high-resolution mass spectrometry.
Leon N, et al.
Journal of Chromatography A, 1258, 55-65 (2012)
Target analysis and retrospective screening of veterinary drugs, ergot alkaloids, plant toxins and other undesirable substances in feed using liquid chromatography-high resolution mass spectrometry.
Leon N, et al.
Talanta, 149, 43-52 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service