Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32551

Supelco

Pefloxacin-d5

VETRANAL®, analytical standard

Synonym(s):

1-Ethyl-d5-6-fluoro-1,4-dihydro-7- (4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, Pefloxacine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D5H15FN3O3
CAS Number:
Molecular Weight:
338.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical
clinical testing
forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])N1C=C(C(O)=O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3

InChI

1S/C17H20FN3O3/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21/h8-10H,3-7H2,1-2H3,(H,23,24)/i1D3,3D2

InChI key

FHFYDNQZQSQIAI-WNWXXORZSA-N

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ki Yong Kim et al.
Journal of hazardous materials, 400, 123235-123235 (2020-09-20)
The concentrations of 14 perfluoroalkyl substances (PFASs) and 46 pharmaceuticals in raw water and drinking water from five drinking water treatment plants were determined to assess removal of the chemicals during treatment. 10 out of 14 PFASs were detected in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service