Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32478

Supelco

Sudan II-(naphthyl-d6)

analytical standard

Synonym(s):

1-(2,4-Dimethyl-phenylazo)naphthalene-d6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D6H10N2O
CAS Number:
Molecular Weight:
282.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.342

pH

10.0±1.0

suitability

no residue for filter test

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c2c(\N=N\c3ccc(C)cc3C)c(O)c([2H])c([2H])c2c1[2H]

InChI

1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3/b20-19+/i3D,4D,5D,6D,8D,10D

InChI key

JBTHDAVBDKKSRW-MUASNCADSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service