Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32437

Supelco

Pyrazoxyfen

PESTANAL®, analytical standard

Synonym(s):

2-[4-(2,4-Dichlorobenzoyl)-1,3-dimethylpyrazol-5-yloxy]acetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Cl2N2O3
CAS Number:
Molecular Weight:
403.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1nn(C)c(OCC(=O)c2ccccc2)c1C(=O)c3ccc(Cl)cc3Cl

InChI

1S/C20H16Cl2N2O3/c1-12-18(19(26)15-9-8-14(21)10-16(15)22)20(24(2)23-12)27-11-17(25)13-6-4-3-5-7-13/h3-10H,11H2,1-2H3

InChI key

FKERUJTUOYLBKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrazoxyfen is used as a pre- or post-emergence herbicide for the weed control in annual and perennial crops.[1]

Application

Pyrazoxyfen may be used as an analytical reference standard for the determination of the analyte in:
  • Shellfish[2] and human serum samples[3] by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction followed by liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) with multiple reaction monitoring (MRM) detection.
  • Food matrices by gas chromatography-mass spectrometry (GC-MS) with MRM detection.[4]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolomics-a robust bioanalytical approach for the discovery of the modes-of-action of pesticides: a review.
Aliferis KA and Jabaji S
Pesticide Biochemistry and Physiology, 100(2), 105-117 (2011)
Compensation of matrix effects in gas chromatography-mass spectrometry analysis of pesticides using a combination of matrix matching and multiple isotopically labeled internal standards.
Tsuchiyama T, et al.
Journal of Chromatography A, 1524(13), 233-245 (2017)
Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography?Tandem Mass Spectrometry.
Shin Y, et al.
Journal of Agricultural and Food Chemistry, 66(13), 3550-3560 (2018)
Analysis of banned veterinary drugs and herbicide residues in shellfish by liquid chromatography-tandem mass spectrometry (LC/MS/MS) and gas chromatography-tandem mass spectrometry (GC/MS/MS).
Chang GR, et al.
Marine Pollution Bulletin, 113(1-2), 579-584 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service