Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32390

Supelco

Ipfencarbazone

PESTANAL®, analytical standard

Synonym(s):

1-(2,4-Dichlorophenyl)-2′,4′-difluoro-1,5-dihydro-N-isopropyl-5-oxo-4H-1,2,4-triazole-4-carboxanilide, 1-(2,4-Dichlorophenyl)-N-(2,4-difluorophenyl)-1,5-dihydro-N-(1-methylethyl)-5-oxo-4H-1,2,4-triazol-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl2F2N4O2
CAS Number:
Molecular Weight:
427.23
Beilstein/REAXYS Number:
14821046
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Product 32390 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(N(C=NN1C2=C(Cl)C=C(Cl)C=C2)C1=O)N(C3=C(F)C=C(F)C=C3)C(C)C

InChI

1S/C18H14Cl2F2N4O2/c1-10(2)25(16-6-4-12(21)8-14(16)22)17(27)24-9-23-26(18(24)28)15-5-3-11(19)7-13(15)20/h3-10H,1-2H3

InChI key

DHYXNIKICPUXJI-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service