Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32381

Supelco

DEET-(diethyl-d10)

PESTANAL®, analytical standard

Synonym(s):

N,N-(Diethyl-d10)-3-methylbenzamide, N,N-(Diethyl-d10)-m-toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10H7NO
CAS Number:
Molecular Weight:
201.33
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(N(C(C([2H])([2H])[2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H])C1=CC(C)=CC=C1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3/i1D3,2D3,4D2,5D2

InChI key

MMOXZBCLCQITDF-IZUSZFKNSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service