Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32320

Sigma-Aldrich

Sodium citrate tribasic dihydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein/REAXYS Number:
6104939
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

agency

USP/NF
reag. ISO
reag. Ph. Eur.

assay

≥99.5%

impurities

≤0.001% total nitrogen (N)
11-13% water (Karl Fischer)

pH

7.5-9.0 (20 °C, 5%)

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
oxalate (C2O42-): ≤200 mg/kg
phosphate (PO43-): ≤20 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Pb: ≤2 mg/kg
Zn: ≤5 mg/kg

suitability

complies for reaction against H2SO4

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

Inchi Key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrocatalytic activity of carbon-supported Pt-Au nanoparticles for methanol electro-oxidation.
Park IS, et al.
Electrochimica Acta, 52(18), 5599-5605 (2007)
Sukdeb Pal et al.
Applied and environmental microbiology, 73(6), 1712-1720 (2007-01-31)
In this work we investigated the antibacterial properties of differently shaped silver nanoparticles against the gram-negative bacterium Escherichia coli, both in liquid systems and on agar plates. Energy-filtering transmission electron microscopy images revealed considerable changes in the cell membranes upon
Toxicity of silver nanoparticles?nanoparticle or silver ion?
Beer C, et al.
Toxicology Letters, 208(3), 286-292 (2012)
Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service