Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32297

Supelco

2-Hydroxy-propoxycarbazone

PESTANAL®, analytical standard

Synonym(s):

2-{{{[4,5-Dihydro-3-(2-hydroxypropoxy)-4-methyl-5-oxo-1H-1,2,4-triazol-1-yl]carbonyl}amino}sulfonyl}benzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O8S
CAS Number:
Molecular Weight:
414.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

O=C(OC)C1=C(S(=O)(NC(N2N=C(OCC(O)C)N(C)C2=O)=O)=O)C=CC=C1

InChI

1S/C15H18N4O8S/c1-9(20)8-27-14-16-19(15(23)18(14)2)13(22)17-28(24,25)11-7-5-4-6-10(11)12(21)26-3/h4-7,9,20H,8H2,1-3H3,(H,17,22)

InChI key

DUXDBOLTMWLLPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service