Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

32276

Aminocyclopyrachlor

PESTANAL®, analytical standard

Synonym(s):

6-Amino-5-chloro-2-cyclopropylpyrimidine-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H8ClN3O2
CAS Number:
Molecular Weight:
213.62
PubChem Substance ID:
UNSPSC Code:
77101502
Beilstein/REAXYS Number:
11329725
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

NC1=C(Cl)C(C(O)=O)=NC(C2CC2)=N1

InChI

1S/C8H8ClN3O2/c9-4-5(8(13)14)11-7(3-1-2-3)12-6(4)10/h3H,1-2H2,(H,13,14)(H2,10,11,12)

InChI key

KWAIHLIXESXTJL-UHFFFAOYSA-N

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service