32244
Tenuazonic acid-(acetyl-13C2), mixture of diastereomers solution
50 μg/mL in acetonitrile (1:1 mixture of (5S,8S) and (5R,8S)-diastereomers), analytical standard
Synonym(s):
(5RS)-3-Acetyl-13C2-1,5-dihydro-4-hydroxy-5-[(1S)-1-methylpropyl]-2H-pyrrol-2-one
About This Item
Recommended Products
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
50 μg/mL in acetonitrile (1:1 mixture of (5S,8S) and (5R,8S)-diastereomers)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
multi-component solution
storage temp.
−20°C
SMILES string
OC1=C([13C]([13CH3])=O)C(NC1[C@@H](C)CC)=O
InChI
1S/C10H15NO3/c1-4-5(2)8-9(13)7(6(3)12)10(14)11-8/h5,8,13H,4H2,1-3H3,(H,11,14)/t5-,8?/m0/s1/i3+1,6+1
InChI key
CEIZFXOZIQNICU-IFOVEIDJSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
35.6 °F
flash_point_c
2 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service