Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32242

Supelco

Terbutaline-(tert-butyl-d9) acetate salt hemihydrate

VETRANAL®, analytical standard

Synonym(s):

5-(2-tert-Butyl-d9-amino-1-hydroxyethyl)benzene-1,3-diol acetate salt hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D9H10NO3 · C2H4O2 · 0.5H2O
Molecular Weight:
303.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

O.CC(O)=O.CC(O)=O.[2H]C([2H])([2H])C(C)(NCC(O)c1cc(O)cc(O)c1)C([2H])([2H])[2H].[2H]C([2H])([2H])C(NCC(O)c2cc(O)cc(O)c2)(C([2H])([2H])[2H])C([2H])([2H])[2H]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hadas Grossman et al.
Reproduction (Cambridge, England) (2019-12-17)
miR-125a-3p, a post-transcription regulator of Fyn kinase, is expressed in mouse pre-ovulatory follicles; its expression within the follicle decreases towards ovulation. Our aim was to follow the synthesis of miR-125a-3p and regulation of its expression in all follicular compartments, focusing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service