Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32172

Supelco

Hydroxyflubendazole

VETRANAL®, analytical standard

Synonym(s):

Methyl N-[6-[(4-fluorophenyl)hydroxymethyl]-1H-benzimidazol-2-yl]carbamate, Reduced flubendazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14FN3O3
CAS Number:
Molecular Weight:
315.30
Beilstein/REAXYS Number:
8856807
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

InChI

1S/C16H14FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8,14,21H,1H3,(H2,18,19,20,22)

InChI key

GONRXGWUUBPRKF-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanny Fri Fombad et al.
Parasites & vectors, 12(1), 14-14 (2019-01-10)
Loiasis, an often-neglected tropical disease, is a threat to the success of lymphatic filariasis and onchocerciasis elimination programmes in rainforest areas of the central and western Africa. Its control and even its elimination might be possible through the use of
Anton Kaufmann et al.
Journal of the American Society for Mass Spectrometry, 28(12), 2705-2715 (2017-09-14)
Targeted residue screening requires the use of reference substances in order to identify potential residues. This becomes a difficult issue when using multi-residue methods capable of analyzing several hundreds of analytes. Therefore, the capability of in silico fragmentation based on
Lukáš Prchal et al.
Parasitology, 142(5), 648-659 (2014-11-07)
The sheep tapeworm Moniezia expansa is very common parasite, which affects ruminants such as sheep, goats as well as other species. The benzimidazole anthelmintics albendazole (ABZ), flubendazole (FLU) and mebendazole (MBZ) are often used to treat the infection. The drug-metabolizing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service