Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32159

Supelco

Isepamicin sulfate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O12 · xH2SO4
CAS Number:
Molecular Weight:
569.60 (free base basis)
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

InChI

1S/C22H43N5O12.H2O4S/c1-22(35)6-36-20(15(33)18(22)26-2)39-17-8(27-19(34)9(28)4-23)3-7(25)16(14(17)32)38-21-13(31)12(30)11(29)10(5-24)37-21;1-5(2,3)4/h7-18,20-21,26,28-33,35H,3-6,23-25H2,1-2H3,(H,27,34);(H2,1,2,3,4)/t7-,8+,9-,10+,11+,12-,13+,14-,15+,16+,17-,18+,20+,21+,22-;/m0./s1

InChI key

DDXRHRXGIWOVDQ-MGAUJLSLSA-N

General description

Isepamicin sulphate is a drug, which belongs to the aminoglycoside class of antibiotics.[1][2]

Application

Isepamicin sulphate may be used as a reference standard in the separation of isepamicin sulphate from structurally related compounds using high performance liquid chromatography (HPLC), coupled with evaporative light scattering detector.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Kong et al.
Frontiers in microbiology, 11, 1718-1718 (2020-08-28)
The emergence of infections caused by bacterial pathogens that are resistant to current antibiotic therapy is a critical healthcare challenge. Aminoglycosides are natural antibiotics with broad spectrum of activity; however, their clinical use is limited due to considerable nephrotoxicity. Moreover
Synthesis of antibiotic-loaded interporous hydroxyapatite blocks by vacuum method and in vitro drug release testing.
Itokazu M, et al.
Biomaterials, 19(7-9), 817-819 (1998)
Determination of isepamicin sulfate and related compounds by high performance liquid chromatography using evaporative light scattering detection.
Vogel R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(3), 405-412 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service