Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32156

Supelco

Hydroxymethylclenbuterol hydrochloride

VETRANAL®, analytical standard

Synonym(s):

2-[2-(4-Amino-3,5-dichlorophenyl)-2-hydroxyethylamino]-2-methyl-1-propanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Cl2N2O2 · HCl
CAS Number:
Molecular Weight:
329.65
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

NC(C(Cl)=C1)=C(Cl)C=C1C(O)CNC(C)(C)CO.Cl

InChI

1S/C12H18Cl2N2O2.ClH/c1-12(2,6-17)16-5-10(18)7-3-8(13)11(15)9(14)4-7;/h3-4,10,16-18H,5-6,15H2,1-2H3;1H

InChI key

INXMPUPHELAAAW-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service