Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32121

Supelco

Sulfapyrazole-(sulfanilamide ring-13C6)

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(3-methyl-1-phenyl-1H-pyrazol-5-yl)benzene-13C6-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C10H16N4O2S
CAS Number:
Molecular Weight:
334.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cc1cc(NS(=O)(=O)[13c]2[13cH][13cH][13c](N)[13cH][13cH]2)n(n1)-c3ccccc3

InChI

1S/C16H16N4O2S/c1-12-11-16(20(18-12)14-5-3-2-4-6-14)19-23(21,22)15-9-7-13(17)8-10-15/h2-11,19H,17H2,1H3/i7+1,8+1,9+1,10+1,13+1,15+1

InChI key

MTERSQYMYBGZTP-LQVUGBGWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service