Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32118

Supelco

Sulfamonomethoxine-(phenyl-13C6) hemihydrate

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(6-methoxy-4-pyrimidinyl)benzene-13C6-sulfonamide hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C5H12N4O3S · 0.5 H2O
CAS Number:
Molecular Weight:
295.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O.COc1cc(NS(=O)(=O)[13c]2[13cH][13cH][13c](N)[13cH][13cH]2)ncn1

General description

Sulfamonomethoxine-(phenyl-13C6) hemihydrate, (CAS:1416768-32-5 (anhydrous)) is a high purity veterinary drug standard used for the determination of the active ingredients of the drug in processed food of animal origin.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service