Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32111

Supelco

Imidocarb-(diethylene-d8) dihydrochloride monohydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D8H12N6O · 2HCl · H2O
Molecular Weight:
447.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.Cl.Cl.[2H]C1([2H])NC(=NC1([2H])[2H])c2cccc(NC(=O)Nc3cccc(c3)C4=NC([2H])([2H])C([2H])([2H])N4)c2

InChI

1S/C19H20N6O.2ClH.H2O/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;;;/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*1H;1H2/i7D2,8D2,9D2,10D2;;;

InChI key

ZZMCPNKUQHIZDQ-YSXHMWEVSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasmine M Aly et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(3), 925-934 (2020-02-13)
NAV 3 is a tumor suppressor of unknown function in leiomyomas. The objective of this study is to assess NAV3 expression and its potential role in human uterine leiomyomas. NAV3 protein expression was examined in patient leiomyoma and patient-matched myometrial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service