Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32092

Supelco

Tecloftalam

PESTANAL®, analytical standard

Synonym(s):

2′,3,3′,4,5,6-Hexachlorophthalanilic acid, 2,3,4,5-Tetrachloro-6-{[(2,3-dichlorophenyl)amino]carbonyl}benzoic acid, 3,4,5,6-Tetrachloro-N-(2,3-dichlorophenyl)phthalamic acid, Techlofthalam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H5Cl6NO3
CAS Number:
Molecular Weight:
447.91
Beilstein/REAXYS Number:
13427114
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C(=O)Nc2cccc(Cl)c2Cl

InChI

1S/C14H5Cl6NO3/c15-4-2-1-3-5(8(4)16)21-13(22)6-7(14(23)24)10(18)12(20)11(19)9(6)17/h1-3H,(H,21,22)(H,23,24)

InChI key

ROZUQUDEWZIBHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of N-(2,3-dichlorophenyl)-3,4,5,6-tetrachlorophthalamic acid (techlofthalam) in paddy soil and rice.
D Kirkpatrick et al.
Journal of agricultural and food chemistry, 29(6), 1149-1153 (1981-11-01)
Biokinetics and metabolism of N-(2,3-dichlorophenyl)-3,4,5,6-tetrachlorophthalamic acid in rats.
D Kirkpatrick et al.
Journal of agricultural and food chemistry, 29(3), 608-614 (1981-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service