Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32031

Supelco

AQUANAL-professional tube test COD

1000-15000 mg/L

Synonym(s):

COD cuvette tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

packaging

pkg of 150 pieces

measuring range

1000-15000 mg/L

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.[1] COD cuvette test is generally used for measuring chemical oxygen demand (COD) in agro-industrial wastes containing natural phenolic compounds.[2]

Analysis Note

oxidation by potassium dichromate in sulfuric acid solution

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 1B - Skin Corr. 1A - STOT RE 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photo-Fenton treatment of water containing natural phenolic pollutants.
Wolfgang G
Chemosphere, 50 (1), 71-78 (2003)
Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service