Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

320153

Sigma-Aldrich

Benzoyl chloride

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCl
CAS Number:
Molecular Weight:
140.57
Beilstein/REAXYS Number:
471389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.88 (vs air)

vapor pressure

1 mmHg ( 32 °C)

product line

ReagentPlus®

assay

99%

autoignition temp.

1056 °F

expl. lim.

4.9 %

refractive index

n20/D 1.553 (lit.)

bp

198 °C (lit.)

mp

−1 °C (lit.)

density

1.211 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H

Inchi Key

PASDCCFISLVPSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High activity of iron containing metal-organic-framework in acylation of p-xylene with benzoyl chloride.
Kurfirtova L, et al.
Catalysis Today, 179(1), 85-90 (2012)
Solid catalyst treated with anion: XVIII. Benzoylation of toluene with benzoyl chloride and benzoic anhydride catalysed by solid superacid of sulfate-supported alumina.
Arata K and Hino M.
Applied Catalysis, 59(1), 197-204 (1990)
Adem Asan et al.
Journal of chromatography. A, 988(1), 145-149 (2003-03-22)
A simple reversed-phase LC method capable of detecting ng/ml quantities of phenolic compounds in water is described. Pre-column derivatization with benzoyl chloride is used for the separation and determination o-cresol, m-cresol, p-cresol, phenol, resorcinol, catechol and hydroquinone in water. The
An unexpected silver triflate-catalyzed reaction of 2-alkynylbenzaldoxime in the presence of benzoyl chloride.
Liu G, et al.
Royal Society of Chemistry Advances, 3(27), 10666-10668 (2013)
Highly efficient catalytic asymmetric acylation of meso-1, 2-diols with benzoyl chloride in the presence of a chirai diamine combined with Et3 N.
Oriyama T, et al.
Tetrahedron Letters, 39(21), 3529-3532 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service