Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31831

Supelco

Kjeldahl Catalyst according to Missouri

tablet, without Hg, Se, Sb, Ti, Pt

Synonym(s):

Nitrogen determination (Missouri)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

form

tablet

suitability

in accordance for function test

application(s)

food and beverages
forensics and toxicology
general analytical

SMILES string

[Na+].[Na+].[Cu++].[O-]S([O-])(=O)=O.[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/Cu.2Na.H2O8S2.H2O4S/c;;;1-9(2,3)7-8-10(4,5)6;1-5(2,3)4/h;;;(H,1,2,3)(H,4,5,6);(H2,1,2,3,4)/q+2;2*+1;;/p-4

InChI key

PIAKSAKPSSOOGC-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Kjeldahl′s method is generally used for the determination of nitrogen in proteins.[1] There are many Kjeldahl catalyst, like copper catalyst Kjeldahl method can be used for the determination of crude protein in feed materials.[2] Mercuric oxide Kjeldahl catalyst is a bit toxic and can be replaced by titanium dioxide, for estimation of total nitrogen content in cereal grains.[3]

Packaging

250 tablets 2.5 g each

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A STUDY OF CONDITIONS FOR KJELDAHL DETERMINATION OF NITROGEN IN PROTEINS DESCRIPTION OF METHODS WITH MERCURY AS CATALYST, AND TITRIMETRIC AND GASOMETRIC MEASUREMENTS OF THE AMMONIA FORMED.
Hiller A
The Journal of Biological Chemistry, 176 (3), 1401-1420 (1948)
The use of titanium dioxide as a catalyst for large-scale kjeldahl determination of the total nitrogen content of cereal grains.
WIlliams PC.
Journal of the Science of Food and Agriculture, 24 (3), 343-348 (1973)
Comparison of LECO FP-228" nitrogen determinator" with AOAC copper catalyst Kjeldahl method for crude protein.
Sweeney RA and Rexroad PR
Journal - Association of Official Analytical Chemists, 70 (6), 1028-1030 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service