Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

31709

Mestranol

VETRANAL®, analytical standard

Synonym(s):

(17α)-3-Methoxy-19-norpregna-1,3,5(10)-trien-20-yn-17-ol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol 3-methyl ether, 17α-Ethynylestradiol 3-methyl ether, 3-Methoxy-17α-ethynylestradiol, NSC 84032

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
EC Number:
200-777-8
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
2625905
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

153-155 °C (lit.)

application(s)

pharmaceutical

format

neat

SMILES string

COc1ccc2C3CC[C@@]4(C)C(CC[C@@]4(O)C#C)C3CCc2c1

InChI

1S/C21H26O2/c1-4-21(22)12-10-19-18-7-5-14-13-15(23-3)6-8-16(14)17(18)9-11-20(19,21)2/h1,6,8,13,17-19,22H,5,7,9-12H2,2-3H3/t17-,18-,19+,20+,21+/m1/s1

InChI key

IMSSROKUHAOUJS-MJCUULBUSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis R Doose et al.
Epilepsia, 44(4), 540-549 (2003-04-12)
To study the pharmacokinetics of a combination oral contraceptive (OC) containing norethindrone and ethinyl estradiol during OC monotherapy, concomitant OC and topiramate (TPM) therapy, and concomitant OC and carbamazepine (CBZ) therapy in order to comparatively evaluate the pharmacokinetic interaction, which
Yuegang Zuo et al.
Journal of chromatography. A, 1148(2), 211-218 (2007-04-03)
A rapid microwave-accelerated derivatization process for the GC-MS analysis of steroid estrogens, estrone (E1), 17beta-estradiol (E2), estriol (E3), 17alpha-ethynylestradiol (EE2) and mestranol (MeEE2), was developed. Under microwave irradiation, the five estrogenic hormones studied were simultaneously derivatized with N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA)+trimethylchlorosilane (TMCS)
A sesquineolignan with a spirodienone structure from Pinus sylvestris L.
Jari Sinkkonen et al.
Angewandte Chemie (International ed. in English), 46(22), 4148-4150 (2007-04-25)
Yuegang Zuo et al.
Chemosphere, 63(9), 1583-1590 (2005-11-26)
17alpha-Ethinylestradiol (EE2), a major constituent of common contraceptive pills, and three other estrogenic hormones, estrone (E1), 17beta-estradiol (E2) and mestranol (MeEE2) have been determined in Acushnet River Estuary seawater using a GC-MS technique. Among three estrogenic compounds detected, EE2 has
Dóra Kovács et al.
Steroids, 77(11), 1075-1085 (2012-05-23)
Regioselective 1,3-dipolar cycloadditions of different aryl nitrile oxides to mestranol were carried out to furnish novel steroidal 17α-isoxazoles in good to excellent yields. Copper(I) was found to be an efficient catalyst, accelerating the intermolecular ring-closures and leading exclusively to 3,5-disubstituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service