Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

31597

Supelco

o-Anisidine

analytical standard

Synonym(s):

2-Aminoanisole, 2-Methoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NH2
CAS Number:
Molecular Weight:
123.15
Beilstein/REAXYS Number:
386210
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.574 (lit.)

bp

225 °C (lit.)

mp

3-6 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COc1ccccc1N

InChI

1S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3

InChI key

VMPITZXILSNTON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Muta. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mazur
The European physical journal. E, Soft matter, 22(1), 67-75 (2007-03-06)
We report on chemical polymerization of two aniline derivatives - 2-methylaniline and 2-methoxyaniline -- in a two-phase water/pentane system. We have found that poly(2-methylaniline) is produced in form of an amorphous material regardless of the fact whether the monomer is
Marie Stiborová et al.
International journal of cancer, 116(5), 667-678 (2005-04-14)
2-methoxyaniline (o-anisidine) is an industrial and environmental pollutant and a bladder carcinogen for rodents. The mechanism of its carcinogenicity was investigated with 2 independent methods, 32P-postlabeling and 14C-labeled o-anisidine, to show that o-anisidine binds covalently to DNA in vitro after
Arnab Dawn et al.
The journal of physical chemistry. B, 110(37), 18291-18298 (2006-09-15)
A novel nano-biocomposite of silver and poly(o-methoxy aniline) (POMA)/DNA hybrid has been prepared by adding DNA solution to an aqueous solution of POMA (emeraldine base, EB) and AgNO(3) mixture. The mixture was aged for 10 days and was freeze-dried to
Xin-Gui Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(18), 4573-4581 (2009-03-20)
The highest Hg-ion adsorbance so far, namely up to 2063 mg g(-1), has been achieved by poly(aniline-co-5-sulfo-2-anisidine) nanosorbents. Sorption of Hg ions occurs mainly by redox and chelation mechanisms (see scheme), but also by ion exchange and physisorption.Poly(aniline (AN)-co-5-sulfo-2-anisidine (SA))
P Santhosh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(7), 1427-1439 (2003-04-26)
UV-visible spectroelectrochemical studies on copolymerization of diphenylamine (DPA) with ortho-methoxy aniline (OMA) were carried out for different feed ratios of DPA and OMA using indium tin oxide (ITO)-coated glass as working electrode. The UV-visible spectra show clear dependencies on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service