Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31595

Supelco

3,5-Dichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein/REAXYS Number:
1906458
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

233 °C (lit.)

mp

65-68 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1cc(Cl)cc(Cl)c1

InChI

1S/C6H4Cl2O/c7-4-1-5(8)3-6(9)2-4/h1-3,9H

InChI key

VPOMSPZBQMDLTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dichlorophenol is a chlorinated phenol mainly used as a pesticide, disinfectant, and as a chemical intermediate in the production of dyes, plastics and pharmaceuticals.

Application

3,5-Dichlorophenol may be used as a reference standard in the determination of the analyte:
  • In the water sample by simultaneous dispersive liquid–liquid microextraction (DLLME) and derivatization combined with gas chromatography-electron-capture detection (GC-ECD) method.
  • In human urine sample by gas chromatography method.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linking mode of action of the model respiratory and photosynthesis uncoupler 3, 5-dichlorophenol to adverse outcomes in Lemna minor.
Xie L, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 197, 98- 108 (2018)
Determination of trace amounts of chlorinated phenols in human urine by gas chromatography.
Edgerton TR, et al.
Analytical Chemistry, 52(11), 1774- 1777 (1980)
Determination of chlorophenols in water samples using simultaneous dispersive liquid--liquid microextraction and derivatization followed by gas chromatography-electron-capture detection.
Fattahi N, et al.
Journal of Chromatography A, 1157(1-2), 23-29 (2007)
Shu-Ling Lin et al.
Analytica chimica acta, 871, 57-65 (2015-04-08)
In this study, methacrylic acid (MAA) was incorporated with alkyl methacrylates to increase the hydrophilicity of the synthesized ethylene dimethacrylate-based (EDMA-based) monoliths for separating polar small molecules by capillary LC analysis. Different alkyl methacrylate-MAA ratios were investigated to prepare a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service