Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31566

Supelco

m-Toluidine

analytical standard

Synonym(s):

3-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein/REAXYS Number:
635944
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.567 (lit.)

bp

203-204 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc(N)c1

InChI

1S/C7H9N/c1-6-3-2-4-7(8)5-6/h2-5H,8H2,1H3

InChI key

JJYPMNFTHPTTDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purnendu Pal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(1-2), 167-171 (2003-12-13)
Menadione (vitamin K(3)) has been shown to form charge transfer complexes with N,N-dimethyl aniline, N,N-dimethyl p-toluidine and N,N-dimethyl m-toluidine in CCl(4) medium. The CT transition energies are well correlated with the ionisation potentials of the anilines. The formation constants of
Gunjan Garg et al.
The journal of physical chemistry. B, 109(4), 1340-1346 (2006-07-21)
The growth behavior of aggregates formed in aqueous solutions of the anionic surfactant sodium dodecyl sulfate (SDS) in the presence of the cationic hydrophobic salts o-toluidine hydrochloride (OTHC) and m-toluidine hydrochloride (MTHC) has been studied by dynamic light scattering (DLS)
N Hada et al.
Mutation research, 493(1-2), 115-126 (2001-08-23)
Norharman, abundantly present in cigarette smoke and cooked foods, is not mutagenic to Salmonella typhimurium strains. However, norharman shows mutagenicity to S. typhimurium TA98 and YG1024 in the presence of S9 mix when coexisting with aromatic amines, including aniline, o-
Metabolism of ortho-, meta-, and para-toluidine in the adult male rat.
K L Cheever et al.
Toxicology and applied pharmacology, 56(3), 361-369 (1980-12-01)
S A Hutcheson et al.
The Journal of chemical physics, 129(7), 074502-074502 (2008-12-03)
Determination of the mechanical response of materials can be fraught with error if rheometer compliance is not properly taken into account. The resulting inaccuracies in the determined mechanical properties of the materials of interest can result in mistakes in material

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service