Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31559

Supelco

Metribuzin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N4OS
CAS Number:
Molecular Weight:
214.29
Beilstein/REAXYS Number:
746650
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CSC1=NN=C(C(=O)N1N)C(C)(C)C

InChI

1S/C8H14N4OS/c1-8(2,3)5-6(13)12(9)7(14-4)11-10-5/h9H2,1-4H3

InChI key

FOXFZRUHNHCZPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomás Undabeytia et al.
Pest management science, 67(3), 271-278 (2011-02-11)
Metribuzin is a widely used herbicide that has been identified as a groundwater contaminant. In this study, slow-release formulations of metribuzin were designed by encapsulating the active ingredient in phosphatidylcholine (PC) vesicles and adsorbing the vesicles onto montmorillonite. The maximum
Neera Singh et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(2), 89-98 (2012-01-19)
This investigation was undertaken to determine the effect of two different fly ashes [Kota and Inderprastha (IP)] amendment on the sorption behavior of metribuzin in three Indian soil types. The IP fly ash was very effective in increasing the metribuzin
Neera Singh et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(2), 108-113 (2013-01-12)
This study reports the effect of fly ash amendment on persistence of metribuzin in three Indian soil types. Fly ash [Inderprastha (IP) and Kota] was amended at 1, 2 and 5% levels. Metribuzin was more persistent in the flooded soils
Lucie Plhalova et al.
TheScientificWorldJournal, 2012, 728189-728189 (2012-07-14)
The aim of this study was to assess the impact of metribuzin in surface waters on fish under experimental conditions. The effects of subchronic exposure to metribuzin on fish growth and the development of histopathological changes in selected organs (gill
Shigemi Kai et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 52(1), 28-33 (2011-03-09)
A method for simultaneous determination of metribuzin (MET) and three metribuzin metabolites in livestock products and seafoods by liquid chromatography-tandem mass spectrometry (LC-MS/MS) was developed. MET and its metabolites were extracted from a sample with acetonitrile, followed by InertSepC18 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service