Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31517

Supelco

Chlorden

PESTANAL®, analytical standard

Synonym(s):

1,7,8,9,10,10-Hexachlorotricyclo[4.3.12,5]deca-3,8-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Cl6
CAS Number:
Molecular Weight:
338.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product 31517 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)[C@@]2(Cl)C3C=CCC3[C@]1(Cl)C2(Cl)Cl

InChI

1S/C10H6Cl6/c11-6-7(12)9(14)5-3-1-2-4(5)8(6,13)10(9,15)16/h1-2,4-5H,3H2/t4?,5?,8-,9+/m0/s1

Inchi Key

XCJXQCUJXDUNDN-DIVXIXLHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of gamma-chlordene in freshwater fish from the Tama River (Japan).
S Horii et al.
Bulletin of environmental contamination and toxicology, 26(2), 254-257 (1981-02-01)
Determination of octanol-water partition coefficients for the major components of technical chlordane.
C D Simpson et al.
Bulletin of environmental contamination and toxicology, 55(1), 149-153 (1995-07-01)
Li Shen et al.
Environmental science & technology, 45(2), 693-699 (2010-12-08)
A chlorinated compound (Chlordene Plus, CP), structurally related to Dechloranes (Dec) 602, 603, 604, and Dechlorane Plus (DP), was identified, and concentrations and spatial trends of Dec 602, 603, 604, CP, and DP in tributary sediments of the Laurentian Great
V J Elia et al.
Environmental research, 32(2), 360-371 (1983-12-01)
The use of municipal wastewater treatment plants for the disposal of industrial wastes creates the potential for the exposure of treatment plant workers to hazardous chemical compounds that may be present in these wastes. Urine from workers and air and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service