Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31408

Sigma-Aldrich

Magnesia sticks

for pearl-tests, package of 25 magnesia sticks

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: magnesium

SMILES string

O=[Mg]

InChI

1S/Mg.O

Inchi Key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas M Tefft et al.
ACS synthetic biology, 8(7), 1590-1600 (2019-06-28)
Microbial electrosynthesis is an emerging technology with the potential to simultaneously store renewably generated energy, fix carbon dioxide, and produce high-value organic compounds. However, limited understanding of the route of electrons into the cell remains an obstacle to developing a
Solaiman Tarafder et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(3), 679-690 (2014-07-22)
The presence of interconnected macro pores allows guided tissue regeneration in tissue engineering scaffolds. However, highly porous scaffolds suffer from having poor mechanical strength. Previously, we showed that microwave sintering could successfully be used to improve mechanical strength of macro
Lin Zhong et al.
Drug development and industrial pharmacy, 40(12), 1660-1669 (2013-10-09)
The amorphous solid dispersions of telmisartan salts were prepared by cogrinding, in presence of alkalizers and polyvinylpyrrolidone (PVPk30). Five alkalizers in this study were MgO, Na2CO3, K2CO3, NaHCO3 and meglumine. In soft mode using a roll mill, the drug could
Tobias C Keller et al.
ChemSusChem, 7(6), 1729-1738 (2014-04-24)
High-silica zeolites have been reported recently as efficient catalysts for liquid- and gas-phase condensation reactions because of the presence of a complementary source of basicity compared to Al-rich basic zeolites. Herein, we describe the controlled generation of these active sites
Kazuhiro Iguchi et al.
Oncology letters, 7(5), 1665-1668 (2014-04-26)
Prostate cancer occurs more frequently among older males and such elderly individuals often have chronic underlying disorders for which various drugs are administered for treatment. The levels of prostate-specific antigen (PSA), a widely used prostate cancer marker, are influenced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service