Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31406

Millipore

DEV Tryptophan Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

meat peptone, 10 g/L
sodium chloride, 5 g/L
DL-tryptophan, 1 g/L

final pH

7.2±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

nonselective for Escherichia coli
nonselective for bacteria (General Media)
nonselective for coliforms

Application

This medium is intended for subcultivation for coliform differentiation, and for indole testing in the bacteriological examination of water.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service