Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31302

Supelco

2-Bromophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OH
CAS Number:
Molecular Weight:
173.01
Beilstein/REAXYS Number:
1905115
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.589 (lit.)

bp

195 °C (lit.)

mp

3-7 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Oc1ccccc1Br

InChI

1S/C6H5BrO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

VADKRMSMGWJZCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

107.6 °F - closed cup

flash_point_c

42 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miho Uchida et al.
Journal of hazardous materials, 101(3), 231-238 (2003-08-26)
2-Bromophenol was reacted in aqueous sodium hydroxide at 200-250 degrees C. The decomposition rate was remarkably faster at 250 degrees C than at 225 or 200 degrees C, and the percentage debromination reached almost 100% in 1M NaOH at 250
Catherine S Evans et al.
Environmental science & technology, 40(9), 3036-3042 (2006-05-25)
The homogeneous, gas-phase oxidative thermal degradation of a 50:50 mixture of 2-bromophenol and 2-chlorophenol was studied in a 1 cm i.d., fused silica flow reactor at a concentration of 88 ppm, with a reaction time of 2.0 s, over a
Christian Eidamshaus et al.
Organic letters, 10(19), 4211-4214 (2008-08-30)
A one-pot synthesis of benzofurans which utilizes a palladium-catalyzed enolate arylation is described. The process demonstrates broad substrate scope and provides differentially substituted benzofurans in moderate to excellent yields. The utility of the method is further demonstrated by the synthesis
Catherine S Evans et al.
Environmental science & technology, 39(7), 2128-2134 (2005-05-06)
The homogeneous, gas-phase oxidative thermal degradation of 2-bromophenol was studied in a 1 cm i.d., fused silica flow reactor at a concentration of 88 ppm, reaction time of 2.0 s, over a temperature range from 300 to 1000 degrees C.
Anusha Jayaraman et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 910, 138-148 (2012-04-24)
This work is focused on the study of the photodegradation of 2-bromophenol under the action of UV light and sunlight. The photodegradation process has been monitored using UV-Vis spectroscopy and High Performance Liquid Chromatography coupled to diode array and mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service