Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31215

Supelco

2-Chloroaniline

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein/REAXYS Number:
606077
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

assay

98-100% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

208-210 °C (lit.)

density

1.213 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1ccccc1Cl

InChI

1S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

AKCRQHGQIJBRMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

221.0 °F - closed cup

flash_point_c

105 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Andini et al.
Journal of hazardous materials, 157(2-3), 599-604 (2008-02-22)
Fuel oil fly ash has been tested as low-cost carbon-based adsorbent of 2-chlorophenol (CP), 2-chloroaniline (CA) and methylene blue (MB) from aqueous solutions. In all the cases the adsorption was found to be of cooperative type. Different adsorption capacities were
Phototransformation of 2-chloroaniline in aqueous solution.
K Othmen et al.
Bulletin of environmental contamination and toxicology, 59(6), 924-927 (1997-12-24)
L Zampori et al.
Journal of hazardous materials, 170(2-3), 1041-1049 (2009-06-16)
The aim of the present work is to investigate the effect of a prolonged leaching test (more than a year) on the microstructure of solidified cementitious wasteforms. A set of four different cement-based monoliths (Ap, Bp, Cp and Dp) was
Dmitry Tsvelikhovsky et al.
Journal of the American Chemical Society, 132(40), 14048-14051 (2010-09-23)
The Pd-catalyzed condensation of 2-bromostyrene and 2-chloroaniline derivatives yields stable diphenylamine intermediates, which are selectively converted to five-, six-, or seven-membered heteroaromatics (indoles, carbazoles, acridines, and dibenzazepines). The selectivity of these intramolecular transformations is uniquely ligand-controlled and offers efficient routes
W K Choy et al.
Chemosphere, 66(11), 2106-2113 (2006-11-23)
Photodecay of o-chloroaniline (o-ClA) in various combinations of UV sources, TiO2, and oxyhalogens was investigated. To improve the conventional photocatalytic process by using UV/TiO2, the addition of oxyhalogens (ClO3(-), BrO3(-) and IO3(-)) into UV/TiO2 system was studied and the effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service