Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31057

Supelco

2,4-D butylglycol ester

PESTANAL®, analytical standard

Synonym(s):

2,4-D butylglycol ester, 2,4-Dichlorophenoxyacetic acid butylglycol ester, 2-Butoxyethyl 2,4-dichlorophenoxyacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl2O4
CAS Number:
Molecular Weight:
321.20
Beilstein/REAXYS Number:
1996617
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOCCOC(=O)COc1ccc(Cl)cc1Cl

InChI

1S/C14H18Cl2O4/c1-2-3-6-18-7-8-19-14(17)10-20-13-5-4-11(15)9-12(13)16/h4-5,9H,2-3,6-8,10H2,1H3

InChI key

ZMWGIGHRZQTQRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stress response of juvenile sockeye salmon (Oncorhynchus nerka) to the butoxyethanol ester of 2,4-dichlorophenoxyacetic acid.
J R McBride et al.
Bulletin of environmental contamination and toxicology, 27(6), 877-884 (1981-12-01)
Acute toxicity to juvenile Pacific salmonids and rainbow trout of butoxyethyl esters of 2,4-D, 2,4-DP and their formulated product: Weedone CB and its carrier.
M T Wan et al.
Bulletin of environmental contamination and toxicology, 45(4), 604-611 (1990-10-01)
J M Charles et al.
Mutation research, 444(1), 227-234 (1999-09-08)
The potential for 2,4-D and seven of its salts and esters to induce cytogenetic abnormalities in mammalian cells in vivo was investigated in the mouse bone marrow micronucleus test. All the test materials were administered to male and female mice
B B Gollapudi et al.
Mutation research, 444(1), 217-225 (1999-09-08)
2,4-dichlorophenoxyacetic acid and its derivatives (collectively known as 2,4-D) are herbicides used to control a wide variety of broadleaf and woody plants. The genetic toxicity of an ester (2,4-D 2-butoxyethylester) and two salts (2,4-D isopropylamine and 2,4-D triisopropanolamine) was investigated
Toxicities of butoxyethanol ester and propylene glycol butyl ether ester formulations of 2,4-dichlorophenoxy acetic acid (2,4-D) to juvenile salmonids.
B J Finlayson et al.
Archives of environmental contamination and toxicology, 14(2), 153-160 (1985-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service