Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31056

Supelco

N-Phenylthiourea

PESTANAL®, analytical standard

Synonym(s):

1-Phenyl-2-thiourea, PTU, Phenylthiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCSNH2
CAS Number:
Molecular Weight:
152.22
Beilstein/REAXYS Number:
907309
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

145-150 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

NC(=S)Nc1ccccc1

InChI

1S/C7H8N2S/c8-7(10)9-6-4-2-1-3-5-6/h1-5H,(H3,8,9,10)

InChI key

FULZLIGZKMKICU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Monier et al.
Journal of hazardous materials, 209-210, 240-249 (2012-01-27)
In this study, cross-linked magnetic chitosan-phenylthiourea (CSTU) resin were prepared and characterized by means of FTIR, (1)H NMR, SEM high-angle X-ray diffraction (XRD), magnetic properties and thermogravimetric analysis (TGA). The prepared resin were used to investigate the adsorption properties of
Warrick J Brewer et al.
Psychiatry research, 199(1), 8-11 (2012-04-17)
Reports suggesting that schizophrenia participants are more likely to be phenylthiocarbamide (PTC) non-tasters when compared to controls have recently been controversial. If supported, a genetic-based phenotypic variation in PTC taster status is implicated, suggesting a greater illness risk for those
Kathleen L Keller
Journal of food science, 77(3), S143-S147 (2012-03-06)
Research suggests that dietary fat is perceived not only by texture, but also by taste. However, the receptors for chemosensory response to fat have not been identified. We report on 2 genes,TAS2R38 and CD36, that may play a role in
Laila Gannoun-Zaki et al.
Antimicrobial agents and chemotherapy, 57(1), 629-632 (2012-11-02)
The mechanism by which the antitubercular drug isoxyl (ISO) inhibits mycolic acid biosynthesis has not yet been reported. We found that point mutations in either the HadA or HadC component of the type II fatty acid synthase (FAS-II) are associated
Seok-Jun Kim et al.
PloS one, 6(9), e25103-e25103 (2011-10-04)
Galectin-3 is known to regulate cancer metastasis. However, the underlying mechanism has not been defined. Through the DNA microarray studies after galectin-3 silencing, we demonstrated here that galectin-3 plays a key role in up-regulating the expressions of protease-activated receptor-1 (PAR-1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service