Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30993

Supelco

myo-2-Inosose

analytical standard

Synonym(s):

2,4,6/3,5-Pentahydroxycyclohexanone, myo-Inosose-2, scyllo-Inosose, Myoinosose, NSC 55547, Scylloinosose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein/REAXYS Number:
2096229
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

assay

≥85.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C6H10O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-5,7-11H/t1?,2-,3+,4+,5-

InChI key

VYEGBDHSGHXOGT-ZLIBEWLCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite of inositol phosphate metabolism, streptomycin biosynthesis, biosynthesis of secondary metabolites, microbial metabolism in diverse environments and biosynthesis of antibiotics.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a gene cluster enabling Lactobacillus casei BL23 to utilize myo-inositol.
Yebra, M.J., et al.
Applied and Environmental Microbiology, 73(12), 3850-3858 (2007)
Probing the promiscuous active site of myo-inositol dehydrogenase using synthetic substrates, homology modeling, and active site modification.
Daniellou , R., et al.
Biochemistry, 46(25), 7469-7477 (2007)
Modulation of amyloid-beta aggregation and toxicity by inosose stereoisomers.
Nitz, M., et al.
FEBS Journal, 275(8), 1663-1674 (2008)
Purification and properties of Bacillus subtilis inositol dehydrogenase.
Ramaley, R., et al.
The Journal of Biological Chemistry, 254(16), 7684-7690 (1979)
The pathway of myo-inositol degradation in Aerobacter aerogenes. Dehydrogenation and dehydration.
T Berman et al.
The Journal of biological chemistry, 241(4), 800-806 (1966-02-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service