30784
Ethyl valerate
analytical standard
Synonym(s):
Ethyl pentanoate
About This Item
grade
analytical standard
Quality Level
assay
≥99.7% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.401 (lit.)
n20/D 1.401
bp
144-145 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCCCC(=O)OCC
InChI
1S/C7H14O2/c1-3-5-6-7(8)9-4-2/h3-6H2,1-2H3
InChI key
ICMAFTSLXCXHRK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service