Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

30698

Supelco

Tiadinil

PESTANAL®, analytical standard

Synonym(s):

5-(3-Chloro-4-methylanilinocarbonyl)-4-methyl-1,2,3-thiadiazole, N-(3-Chloro-4-methylphenyl)-4-methyl-1,2,3-thiadiazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10ClN3OS
CAS Number:
Molecular Weight:
267.73
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

111-113 °C

application(s)

agriculture
environmental

format

neat

InChI

1S/C11H10ClN3OS/c1-6-3-4-8(5-9(6)12)13-11(16)10-7(2)14-15-17-10/h3-5H,1-2H3,(H,13,16)

InChI key

VJQYLJSMBWXGDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tiadinil may be used as a reference standard for the determination of the analyte in fruits/vegetables,[1] cooked wheat flour and polished rice[2] and human serum[3] by various chromatography techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue analysis of 47 pesticides in cooked wheat flour and polished rice by liquid chromatography with tandem mass spectrometry.
Lee SJ, et al.
Biomedical Chromatography, 23(4), 434-442 (2009)
Simultaneous determination of sixteen amide fungicides in vegetables and fruits by dispersive solid phase extraction and liquid chromatography-tandem mass spectrometry.
Wu YL, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 989, 11-20 (2015)
Validation of a Multiresidue Analysis Method for 379 Pesticides in Human Serum Using Liquid Chromatography?Tandem Mass Spectrometry.
Shin Y, et al.
Journal of Agricultural and Food Chemistry, 66(13), 3550-3560 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service