Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30638

Supelco

Petunidin 3-O-β-D-glucoside chloride

analytical standard

Synonym(s):

Petunidin 3-O-β-D-glucopyranoside, Petunidin 3-glucoside, Petunidin 3-glucoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23ClO12
CAS Number:
Molecular Weight:
514.86
UNSPSC Code:
12352200
NACRES:
NA.24
Product 30638 is not currently sold in your country. Contact Technical Service

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C22H22O12.ClH/c1-31-14-3-8(2-12(26)17(14)27)21-15(6-10-11(25)4-9(24)5-13(10)32-21)33-22-20(30)19(29)18(28)16(7-23)34-22;/h2-6,16,18-20,22-23,28-30H,7H2,1H3,(H3-,24,25,26,27);1H/t16-,18-,19+,20-,22-;/m1./s1

InChI key

HBKZHMZCXXQMOX-YATQZQGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aline Macedo Dantas et al.
Food chemistry, 274, 202-214 (2018-10-31)
The bioaccessibility of phenolics and antioxidant activity were determined in açaí, cupuaçu, blackberry, blueberry, jabuticaba, raspberry, cajá and soursop frozen pulps (FPs) using a digestion model coupled with a simulated intestinal barrier. Cyanidin 3-glucoside (6.56%) and pelargonidin 3-glucoside (28.33%) were
Jang Hoon Kim et al.
Bioorganic chemistry, 87, 803-809 (2019-04-13)
Eight anthocyanidin derivatives (1-8) were evaluated as potential inhibitors of the catalysis of α-glucosidase. Among them, compounds 4 and 8 had the highest levels of inhibitory activity at 100 μM (IC50 values of 14.4 ± 0.1 and 29.7 ± 1.2 μM) and acted in a dose-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service